Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Amylase Reagent, Infinity Amylase EPS 2 x 125mL

Catalog No. TR25421 Shop All Thermo Scientific Products
Change view
Click to view available options
Infinity Amylase EPS 2 x 125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
TR25421 Infinity Amylase EPS 2 x 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TR25421
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Amylase Reagent is suitable for manual use or on automated clinical chemistry analyzers.

Determine quantity of amylase in serum or urine in vitro with Thermo Scientific™ Amylase Reagent, suitable for manual use or on automated clinical chemistry analyzers.

Amylase is most frequently measured in the diagnosis of acute pancreatitis when serum levels may be grossly elevated. Hyperamylasemia is associated with, but not limited to, other acute abdominal disorders, biliary tract disease, diabetic ketoacidosis, severe glomerular dysfunction, salivary gland disorders, ruptured ectopic pregnancy and macroamylasemia. Applications for a wide range of automated analyzers are available on request.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Description Infinity Amylase EPS
CE Marker Yes
Linearity 10 to 2000U/L
Methods Compatibility Kinetic
Quantity 2 x 125 mL
Stability Stable Liquid
State at Room Temperature Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.