Learn More
Enzo Life Sciences Amyloid beta Recombinant monoclonal antibody (WO2) (Mouse IgG2a kappa) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS3860100
Encompass_Preferred
Clone: WO2. Isotype: IgG2aκ. Immunogen: Synthetic peptides, corresponding to aa 1-42 of human beta amyloid, conjugated to KLH. Species reactivity: Human. Specificity: Recognizes aa 4-10 of human Amyloid β. Applications: ELISA, IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2913