Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Amylcinnamyl alcohol, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012302 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2302 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2302 Supplier MilliporeSigma Supelco Supplier No. 87894500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 101-85-9
Density 0.952 g/mL at 25°C; 0.966 g/mL (at 25°C)
Flash Point >110°C
Physical Form Neat
Linear Formula C6H5 CH=C[(CH2)4 CH3 ]CH2 OH
MDL Number MFCD00022004
Boiling Point 141°C to 143°C (5 mmHg)
Synonym 2-Benzylidene-1-heptanol
Type Analytical Standard
Quantity 500 mg
Formula Weight 204.31
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.