Learn More
AnaSpec Amylin (1-37), Rat, Mouse, Synthetic Peptide, Research Use Only, 0.5 mg
Supplier:  AnaSpec AS6025305
Encompass
Amylin (1-37), rat, mouse is a synthetic peptide crucial for research on amylin's physiological roles without amyloid fibril formation. It differs from human amylin by six amino acids and serves as a potent tool for understanding islet amyloid polypeptide functions. This peptide supports various laboratory applications, ensuring quality with a purity of ≥95%.
Catalog No. NC1829082