Learn More
Medchemexpress LLC Amylin, amide, human | 122384-88-7 | 98.3% | 3903.28 | 1 MG
SDP
Catalog No. 5000425204
Encompass_Preferred
Amylin, amide, human is a 37-amino acid polypeptide and a pancreatic hormone cosecreted with insulin. It plays unique roles in metabolism and glucose homeostasis, inhibiting glucagon secretion, delaying gastric emptying, and acting as a satiety agent. This product is for research use only and not sold to patients.
  • Inhibits glucagon secretion
  • Delays gastric emptying
  • Acts as a satiety agent
  • Stimulates human amylin receptor CTR1 and CTR2 in MCF-7 cells
  • Results in cAMP production with an EC50 of 35.2±7.5 nM in MCF-7 cells
  • Subcutaneous injection in Swiss male mice showed a half-time of 23 min
Catalog No. 50-004-25204 Supplier Medchemexpress LLC Supplier No. HYP10701MG
Amylin, amide, human is a 37-amino acid polypeptide and a pancreatic hormone cosecreted with insulin. It plays unique roles in metabolism and glucose homeostasis, inhibiting glucagon secretion, delaying gastric emptying, and acting as a satiety agent. This product is for research use only and not sold to patients.
  • Inhibits glucagon secretion
  • Delays gastric emptying
  • Acts as a satiety agent
  • Stimulates human amylin receptor CTR1 and CTR2 in MCF-7 cells
  • Results in cAMP production with an EC50 of 35.2±7.5 nM in MCF-7 cells
  • Subcutaneous injection in Swiss male mice showed a half-time of 23 min