Learn More
Medchemexpress LLC Amylin, amide, human TFA | 98.8% | 4017.30 | 1 MG
SDP
Catalog No. 5000425014
Encompass_Preferred
Amylin, amide, human TFA is a 37-amino acid polypeptide and a pancreatic hormone that is cosecreted with insulin. It plays unique roles in metabolism and glucose homeostasis by inhibiting glucagon secretion, delaying gastric emptying, and acting as a satiety agent. This product is for research use only and not sold to patients.
  • Inhibits glucagon secretion
  • Delays gastric emptying
  • Acts as a satiety agent
  • Stimulates human amylin receptor CTR1 and CTR2 in MCF-7 cells with an EC50 of 35.2 ± 7.5 nM
  • Exhibits a half-time of 23 minutes in Swiss male mice after subcutaneous injection
Catalog No. 50-004-25014 Supplier Medchemexpress LLC Supplier No. HYP1070A1MG
Amylin, amide, human TFA is a 37-amino acid polypeptide and a pancreatic hormone that is cosecreted with insulin. It plays unique roles in metabolism and glucose homeostasis by inhibiting glucagon secretion, delaying gastric emptying, and acting as a satiety agent. This product is for research use only and not sold to patients.
  • Inhibits glucagon secretion
  • Delays gastric emptying
  • Acts as a satiety agent
  • Stimulates human amylin receptor CTR1 and CTR2 in MCF-7 cells with an EC50 of 35.2 ± 7.5 nM
  • Exhibits a half-time of 23 minutes in Swiss male mice after subcutaneous injection