Learn More
CPC Scientific H-Ala-Thr-Gln-Arg-Leu-Ala-Asn-Phe-Leu-Val-His-Ser-Ser-Asn-Asn-Phe-Gly-Ala-Ile-Leu-Ser-Ser-Thr-Asn-Val-Gly-Ser-Asn-Thr-Tyr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific AMYN003B
Encompass
Amylin (8-37) is a truncated analog of native Amylin that selectively inhibits insulin-related glucose uptake and glycogen deposition in muscle tissue. The peptide has a systemic increase in insulin sensitivity and has shown to reduce basal insulin levels in both normal and hGH-induced insulin-resistant rats.ONE-LETTER SEQUENCE: ATQRLANFLVHSSNNFGAILSSTNVGSNTYMOLECULAR FORMULA: C138H215N41O46MOLECULAR WEIGHT:3184.45STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [135702-23-7], SYNONYMS: Amylin (8-37) (human)RESEARCH AREA: DiabetesREFERENCES: Deems, R.O. et al. Biochem. Biophys. Res. Commun., 181, 116 (1991); Kaygisiz, Z. et al. Acta Physiol. Hung. 90, 133 (2003); B.Konarkowska et al., FEBS J., 272, 4949 (2005)
Catalog No. 50-210-8757