Learn More
Genscript Corporation Amylin, human, amide
Supplier:  Genscript Corporation RP112780.5
Encompass
Amylin, a 37-amino acid polypeptide that is structurally related to calcitonin, is secreted from the B cells of the pancreas. Amylin has anoretic effects in rats. Amylin may be responsible for the etiology of insulin resistance of type II diabetes mellitus through its modulation of peripheral effects of insulin. Amylin blocks the activation of glycogen synthase by insulin.
Catalog No. 50-275-069