Learn More
Abcam Amylin (human), Endogenous peptide, 2500UG
Catalog No. 502706257
Encompass_Preferred
MW 3903.3 Da, Purity >95%. Amylin (human), Endogenous peptide. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6257 Supplier Abcam Supplier No. AB1423982500UG
MW 3903.3 Da, Purity >95%. Amylin (human), Endogenous peptide. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement