Learn More
CPC Scientific H-Ala-Thr-Gln-Arg-Leu-Ala-Asn-Phe-Leu-Val-Arg-Ser-Ser-Asn-Asn-Leu-Gly-Pro-Val-Leu-Pro-Pro-Thr-Asn-Val-Gly-Ser-Asn-Thr-Tyr-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific AMYN005B
Encompass
An amylin receptor antagonist that attenuates the ANP release in streptozotocin-treated rats.ONE-LETTER SEQUENCE: ATQRLANFLVRSSNNLGPVLPPTNVGSNTY-NH2MOLECULAR FORMULA: C140H227N43O43MOLECULAR WEIGHT:3200.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [135702-23-7], RESEARCH AREA: DiabetesREFERENCES: R.O. Deems et al., BBRC, 181, 116 (1991)
Catalog No. 50-210-8759