Learn More
Medchemexpress LLC Amylmetacresol | 1300-94-3 | ≥98.0% | 500 MG
SDP
Catalog No. 5000232319
Encompass_Preferred
Amylmetacresol is a phenolic antiseptic supplied as an analytical reference standard for research and assay development. It exhibits topical antibacterial and antiviral activity and is commonly used in studies of throat actives and antiviral screening. CAS 1300-94-3; molecular formula C12H18O; molecular weight 178.28 g/mol.
  • Phenolic antiseptic with antibacterial and antiviral activity.
  • Suitable as an analytical reference standard for assay and method development.
  • Low molecular weight (178.28 g/mol) and simple chemical formula (C12H18O).
  • Packaged in small reference quantities for analytical use.
  • Used in research on topical throat antiseptics and antiviral agents.
Catalog No. 50-002-32319 Supplier Medchemexpress LLC Supplier No. HY121527500MG
Amylmetacresol is a phenolic antiseptic supplied as an analytical reference standard for research and assay development. It exhibits topical antibacterial and antiviral activity and is commonly used in studies of throat actives and antiviral screening. CAS 1300-94-3; molecular formula C12H18O; molecular weight 178.28 g/mol.
  • Phenolic antiseptic with antibacterial and antiviral activity.
  • Suitable as an analytical reference standard for assay and method development.
  • Low molecular weight (178.28 g/mol) and simple chemical formula (C12H18O).
  • Packaged in small reference quantities for analytical use.
  • Used in research on topical throat antiseptics and antiviral agents.