Learn More
Cayman Chemical Amylmetacresol, Purity: ≥98%, CAS: 1300-94-3, 1 g
SDP
Catalog No. 502978646
Encompass_Preferred
A topical antiseptic and analgesic agent; inhibits depolarization-induced whole-cell sodium currents in HEK293 cells expressing human Na
Catalog No. 50-297-8646 Supplier Cayman Chemical Supplier No. 402901 G
A topical antiseptic and analgesic agent; inhibits depolarization-induced whole-cell sodium currents in HEK293 cells expressing human Na