Learn More
Cayman Chemical Amylmetacresol, Purity: ≥98%, CAS: 1300-94-3, 5 g
SDP
Catalog No. 502978475
Encompass_Preferred
A topical antiseptic and analgesic agent; inhibits depolarization-induced whole-cell sodium currents in HEK293 cells expressing human Na
Catalog No. 50-297-8475 Supplier Cayman Chemical Supplier No. 402905 G
A topical antiseptic and analgesic agent; inhibits depolarization-induced whole-cell sodium currents in HEK293 cells expressing human Na