Learn More
Sigma Aldrich Fine Chemicals Biosciences Amyloglucosidase from Aspe
Catalog No. 502906557
Encompass
Amyloglucosidase a 1 4 -D-glucan glucohydrolase is a disaccharidase-type glucosidase. It is synthesized by several Aspergillus genus species. This exo-enzyme is one of the major industrial enzymes. The stability of amyloglucosidase can be increased by immobilization.
Catalog No. 50-290-6557 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11202367001
Amyloglucosidase a 1 4 -D-glucan glucohydrolase is a disaccharidase-type glucosidase. It is synthesized by several Aspergillus genus species. This exo-enzyme is one of the major industrial enzymes. The stability of amyloglucosidase can be increased by immobilization.