Learn More
AnaSpec AMYLOID (1-40) N-TERMINAL
Catalog No. 50850244
Encompass
Beta-Amyloid (1-40), Human[DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVV] ; MW 4329.9; (0.5 mg) AB (1-40) together with AB (1-42) are two major C-terminal variants of the AB protein constituting the majority of ABs. These undergo post-secretory aggregation and deposition in the Alzheimer’s disease brain.
Catalog No. 50-850-244 Supplier AnaSpec Supplier No. AS24235
Beta-Amyloid (1-40), Human[DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVV] ; MW 4329.9; (0.5 mg) AB (1-40) together with AB (1-42) are two major C-terminal variants of the AB protein constituting the majority of ABs. These undergo post-secretory aggregation and deposition in the Alzheimer’s disease brain.