Learn More
Cayman Chemical AmyloId-1-42 PeptIdetrI 500ug
SDP
Supplier:  Cayman Chemical 20574500 UG
Encompass_Preferred
A 42-amino acid protein fragment of amyloid-β; found in amyloid plaques in postmortem cerebral cortex from patients with Alzheimer’s disease; aggregation results in the formation of neurotoxic fibrils or globular oligomers; accumulates in the brain of many transgenic mouse models of Alzheimer’s disease and, in many models, the onset of amyloid deposition positively correlates with deficits in spatial learning and memory
Catalog No. 50-298-2797