Learn More
Medchemexpress LLC HY-P1388A 1mg Medchemexpress, β-Amyloid (1-42), rat (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1388A1MG
Encompass_Preferred
Medchemexpress, HY-P1388A 1mg β-Amyloid (1-42), rat (TFA) CAS: β-Amyloid (1-42), rat TFA is a 42-aa peptide, shows cytotoxic effect on acute hippocampal slices, and used in the research of Alzheimer's disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4195