Learn More
Medchemexpress LLC HY-P1051A 1mg Medchemexpress, β-amyloid (12-28) (TFA) CAS: Purity:>98%
Supplier: Medchemexpress LLC HYP1051A1MG
Medchemexpress, HY-P1051A 1mg β-amyloid (12-28) (TFA) CAS: β-amyloid (12-28) TFA is a peptide fragment of β-amyloid protein (β1-42). β1–42 a 42 amino acid protein , is the major component of senile plaque cores. β-amyloid (12-28) shows aggregation properties. β-amyloid (12-28) has the potential for Alzheimer’s disease research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.