Learn More
Diagnostic Biosystems AMYLOID Amyloid Stain Kit (Congo Red)
Catalog No. 502407412
Encompass_Preferred
The Amyloid Stain Kit (Congo Red) is intended for use in the histological visualization of amyloid in tissue sections. Examination under a polarizing microscope results in green birefringence of amyloid.
Catalog No. 50-240-7412 Supplier Diagnostic Biosystems Supplier No. KT008
The Amyloid Stain Kit (Congo Red) is intended for use in the histological visualization of amyloid in tissue sections. Examination under a polarizing microscope results in green birefringence of amyloid.