Learn More
AnaSpec Beta - Amyloid (1 - 42), Human 1mg
Supplier:  AnaSpec AS20276
Encompass
Aß (1-42), a major component of amyloid plaques, accumulates in neurons of Alzheimer’s disease brains. Biochemical analysis of the amyloid peptides isolated from Alzheimer’s disease brain indicates that Aß (1-42) is the principal species associated with senile plaque amyloids, while Aß (1-40) is more abundant in cerebrovascular amyloid deposit. MW 4514.1; Purity % Peak Area By HPLC ≥ 95%; Storage -20°C
Catalog No. 50-824-533