Learn More
AnaSpec AMYLOID ?-AMYLOID (1-42)
Supplier:  AnaSpec AS24224
Encompass
Beta-Amyloid (1-42), Human[DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA] ; MW 4514.1; (0.5 mg) AB (1-42), a major component of amyloid plaques, accumulates in neurons of Alzheimer’s disease brains. Biochemical analysis of the amyloid peptides isolated from Alzheimer’s disease brain indicates that AB (1-42) is the principal species associated with senile plaque amyloids, while AB (1-40) is more abundant in cerebrovascular amyloid deposit.
Catalog No. 50-824-486