Learn More
Enzo Life Sciences Amyloid beta A4 (CT, 1-42) monoclonal antibody (8G7) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADI905804100
Encompass_Preferred
Clone: 8G7. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 1-42 of amyloid β A4. UniProt: P05067. Species Reactivity: Human. Crossreactivity: Does not cross react with β A4 (1-40). Applications: ELISA, ICC, WB. Recommended dilutions: ELISA (0.05µg/ml). Immunocytochemistry (0.1-1.0µg/ml). Western Blot (1µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. NC1750185