Learn More
Enzo Life Sciences Amyloid beta A4 (CT, 40/42) monoclonal antibody (9F1) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905803100
Encompass_Preferred
Clone: 9F1. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 1-42 amyloid β A4. UniProt ID: P05067. Species reactivity: Human. Creossreactivity: Cross-reacts with APP. Applications: ELISA, ICC, WB. Recommended Dilutions: ELISA (0.05µg/ml) Immunocytochemistry (0.1-1.0µg/ml) Western Blot (1µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-7067