Learn More
CPC Scientific H-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific AMYD046B
Encompass
beta-Secretase Substrate II used for assaying beta-secretase activity and corresponds to the 'Swedish' Lys-Met/Asn-Leu mutation of amyloid precursor protein (APP) beta-secretase cleavage site. ONE-LETTER SEQUENCE: SEVNLDAEFRMOLECULAR FORMULA: C50H78N14O19MOLECULAR WEIGHT:1179.3STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [186142-28-9], RESEARCH AREA: Alzheimer's DiseaseREFERENCES: F. Grüninger-Leitch, et al., Nat. Biotechnol. 18, 66 (2000); S. Sinha, et al., Nature 402, 537 (1999); R. Vasser, et al., Science 286, 735 (1999); R. Yan, et al., Nature 402, 533 (1999)
Catalog No. 50-210-8704