Learn More
Bachem Amyloid Beta Protein (1-38) Trifluoroacetate
SDP
Supplier:  Bachem 4030204.1000
Encompass_Preferred
Amyloid Beta Protein (1-38) Trifluoroacetate, H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-OH, 131438-74-9, C184H277N51O56S, Mr 4131.60, 1mg. Like Ab (25-35), the Ab fragment (1-38) destabilizes calcium homeostasis and renders human cortical neurones vulnerable to environmental insults.
Catalog No. 50-259-675