Learn More
Bachem Amyloid Beta Protein (25-35) Trifluoroacetate
SDP
Supplier:  Bachem 4030205.0001
Encompass_Preferred
Amyloid Beta Protein (25-35) Trifluoroacetate, H-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-OH, 131602-53-4, C45H81N13O14S, Mr 1060.28, 1mg. The biologically active domain of the amyloid b-protein for neurotrophic and neurotoxic effects is located in the b (25-35) sequence GSNKGAIIGLM, which is homologous to a region in peptides of the tachykinin family.
Catalog No. 50-259-474