Learn More
Bachem Amyloid Beta Protein (35-25) Trifluoroacetate
SDP
Supplier:  Bachem 4030076.0001
Encompass_Preferred
Amyloid Beta Protein (35-25) Trifluoroacetate, H-Met-Leu-Gly-Ile-Ile-Ala-Gly-Lys-Asn-Ser-Gly-OH, 147740-73-6, C45H81N13O14S, Mr 1060.28, 1mg. b-Amyloid (35-25). The reverse sequence of Ab 25-35 is used as inactive control.
Catalog No. 50-259-673