Learn More
Newcomer Supply Amyloid, Bennhold Congo Red Stain Kit, 250 ml kit
SDP
Catalog No. NC2102255
Encompass
Newcomer Supply Amyloid, Bennhold Congo Red Stain Kit procedure, with included microwave modification, is used for identifying the extraneous protein deposits in amyloidosis. The use of polarizing lenses is an essential technique for visualizing amyloid positive areas and/or to confirm negativity.
This item is not returnable. View return policy
Catalog No. NC2102255 Supplier Newcomer Supply Supplier No. 9103A
Newcomer Supply Amyloid, Bennhold Congo Red Stain Kit procedure, with included microwave modification, is used for identifying the extraneous protein deposits in amyloidosis. The use of polarizing lenses is an essential technique for visualizing amyloid positive areas and/or to confirm negativity.