Learn More
IBL America Amyloid Beta (82E1-specific) Aβ Oligomers
Supplier:  IBL America 27725
Encompass_Preferred
Solid phase sandwich ELISA for the determination of human A molecules which bind to anti-human A (N) (82E1) mouse MoAb with two or more epitopes (oligomers, polymers with protein etc.) in serum, EDTA-plasma and brain tissue extract. For research use only, not for use in diagnostic procedures.
Catalog No. 50-136-6532