Learn More
CPC Scientific H-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific AMYD051A
Encompass
This peptide is the biologically active domain of the amyloid beta-protein for neurotrophic and neurotoxic effects, which is homologous to a region in peptides of the tachykinin family. ONE-LETTER SEQUENCE: GSNKGAIIGLMMOLECULAR FORMULA: C45H81N13O14S1MOLECULAR WEIGHT:1060.28STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [131602-53-4], RESEARCH AREA: Alzheimer's DiseaseREFERENCES: B.A.Yankner et al., Science, 250, 279 (1990); A.Klegeris et al., Biochem. Biophys. Res. Commun., 199, 984 (1994); C.J.Pike et al., Neuroscience, 63, 517 (1994); C.J.Pike et al., J. Neurochem., 64, 253 (1995); J.H.Ippel et al., Proc. Natl. Acad. Sci. USA, 99, 8648 (2002); M.E.Clementi et al., FEBS Lett., 579, 2913 (2005); F.Misiti et al., Neurochem. Int., 46, 575 (2005); G.Wei and J.-E.Shea, Biophys. J., 91, 1638 (2006); D.Buntup et al., Neurochem. Res., 33, 248 (2008); I.D.Limon et al., Neurosci. Res., 63, 129 (2009); L.Millucci et al., Curr. Protein Pept. Sci., 11, 54 (2010); H.-H.G.Tsai et al., P
Catalog No. 50-210-8712