Learn More
Sigma Aldrich Fine Chemicals Biosciences Amyloid beta-Protein fragm1MG
Catalog No. 502898840
Encompass
Amyloid -Protein fragment 25-35 (A25-35) is derived from the amyloid- protein.amyloid- protein which is mapped to human chromosome 21q21. A25-35 lacks the N-terminal domain and the metal binding site and is majorly generated by proteolytic cleavage of A(1A40) peptides. It has a -sheet and -turn structure.
Catalog No. 50-289-8840 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A45591MG
Amyloid -Protein fragment 25-35 (A25-35) is derived from the amyloid- protein.amyloid- protein which is mapped to human chromosome 21q21. A25-35 lacks the N-terminal domain and the metal binding site and is majorly generated by proteolytic cleavage of A(1A40) peptides. It has a -sheet and -turn structure.