Learn More
Sigma Aldrich Fine Chemicals Biosciences Amyloid beta Protein fragm1MG
Catalog No. 502898216
Encompass
Amyloid Protein fragment 1-40 (A40) is derived from the amyloid- protein (A) which is mapped to human chromosome 21q21.3. A40 is predominantly present in the vascular amyloid deposits. A40 comprises of C-terminal membrane insertion domain. It shows structural transition from random coil to a helical structure in a water-micelle medium.
Catalog No. 50-289-8216 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A1075.1MG
Amyloid Protein fragment 1-40 (A40) is derived from the amyloid- protein (A) which is mapped to human chromosome 21q21.3. A40 is predominantly present in the vascular amyloid deposits. A40 comprises of C-terminal membrane insertion domain. It shows structural transition from random coil to a helical structure in a water-micelle medium.