Learn More
CPC Scientific Glp-Ala-Ser-Asn-Cys-Phe-Ala-Ile-Arg-His-Phe-Glu-Asn-Lys-Phe-Ala-Val-Glu-Thr-Leu-Ile-Cys-Ser-Arg-Thr-Val-Lys-Lys-Asn-Ile-Ile-Glu-Glu-Asn-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific AMYD038A
Encompass
Peptide encoded by a novel gene, BRI (located on chromosome 13), and deposited as amyloid fibrils causing neuronal dysfunction and dementia.ONE-LETTER SEQUENCE: Glp-ASNCFAIRHFENKFAVETLICSRTVKKNIIEEN MOLECULAR FORMULA: C173H275N49O52S2MOLECULAR WEIGHT:3937.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [321853-88-7], SYNONYMS: Amyloid Bri Protein (1-34) (reduced)RESEARCH AREA: Alzheimer's DiseaseREFERENCES: R. Vidal et al., Nature, 399, 776 (1999)
Catalog No. 50-210-8697