Learn More
Newcomer Supply Amyloid, Animal Control Slides, 10/set
SDP
Catalog No. NC0366362
Encompass
Newcomer Supply Amyloid, Animal Control Slides are for the positive histochemical staining of extraneous protein deposits in amyloidosis. Amyloid, Animal Control Slides contain sections of positive staining animal organism.
Catalog No. NC0366362 Supplier Newcomer Supply Supplier No. 4031A
Newcomer Supply Amyloid, Animal Control Slides are for the positive histochemical staining of extraneous protein deposits in amyloidosis. Amyloid, Animal Control Slides contain sections of positive staining animal organism.