Learn More
CPC Scientific Glp-Ala-Ser-Asn-Cys-Phe-Ala-Ile-Arg-His-Phe-Glu-Asn-Lys-Phe-Ala-Val-Glu-Thr-Leu-Ile-Cys-Phe-Asn-Leu-Phe-Leu-Asn-Ser-Gln-Glu-Lys-His-Tyr-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific AMYD039A
Encompass
ONE-LETTER SEQUENCE: Glp-ASNCFAIRHFENKFAVETLICFNLFLNSQEKHY (C5&C22 bridge)MOLECULAR FORMULA: C185H268N48O51S2MOLECULAR WEIGHT:4044.59STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1802082-60-5], RESEARCH AREA: Alzheimer's Disease
Catalog No. 50-210-8699