Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amyloid P-IN-1 | 1819986-22-5 | MFCD30489728 | 99.6% | 656.68 g/mol | C30H44N2O14 | 10 MG
SDP

Catalog No. 5000294851
Encompass_Preferred

Amyloid P-IN-1 is a research-grade small-molecule inhibitor used to study depletion of serum amyloid P component (SAP) in models of amyloidosis, Alzheimer's disease, type 2 diabetes mellitus, and osteoarthritis. It is supplied for laboratory research use only and is not intended for clinical use or human administration.

  • High purity: 99.6% by assay.
  • Molecular weight: 656.68 g/mol.
  • Chemical formula: C30H44N2O14.
  • Available as milligram quantities or as a 10 mM solution in DMSO.
  • Solid appearance: white to off-white powder.
  • Storage recommendations for powder and solutions to maintain stability.

Catalog No. 50-002-94851 Supplier Medchemexpress LLC Supplier No. HY1977110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amyloid P-IN-1 is a research-grade small-molecule inhibitor used to study depletion of serum amyloid P component (SAP) in models of amyloidosis, Alzheimer's disease, type 2 diabetes mellitus, and osteoarthritis. It is supplied for laboratory research use only and is not intended for clinical use or human administration.

  • High purity: 99.6% by assay.
  • Molecular weight: 656.68 g/mol.
  • Chemical formula: C30H44N2O14.
  • Available as milligram quantities or as a 10 mM solution in DMSO.
  • Solid appearance: white to off-white powder.
  • Storage recommendations for powder and solutions to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.