Learn More
TARGETMOL CHEMICALS INC AMYLOID P-IN-1 5MG
SDP
Catalog No. 502581302
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. amyloid P-IN-1 can be used in studies about diseases with depletion of serum amyloid P components such as Alzheimer's disease amyloidosis osteoarthritis and type 2 diabetes mellitus. purity: 98%
Catalog No. 50-258-1302 Supplier TARGETMOL CHEMICALS INC Supplier No. T142835MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. amyloid P-IN-1 can be used in studies about diseases with depletion of serum amyloid P components such as Alzheimer's disease amyloidosis osteoarthritis and type 2 diabetes mellitus. purity: 98%