Learn More
Apexbio Technology LLC Amyloid Precursor C-Terminal Peptide 5mg
SDP
Catalog No. 5000095787
Encompass_Preferred
Amyloid Precursor C-Terminal Peptide is a peptide fragment derived from Amyloid Precursor Protein It is designed to model the proteolytic cleavage process of APP which yields -amyloid peptide (A ) associated with amyloid plaques in Alzheimer s disease pathology Amyloid Precursor C-Terminal Peptide influences neuronal functions such as synaptogenesis neural plasticity and iron export regulation Its cleavage product A induces astrocyte proliferation via MAPK and NF- B signaling pathways contributing to neurodegeneration Additionally involvement in inflammation and tissue damage has been reported in animal arthritis models Based on these characteristics Amyloid Precursor C-Terminal Peptide holds research potential in studying A formation neurodegeneration mechanisms and inflammation-associated tissue injury
Catalog No. 50-000-95787 Supplier Apexbio Technology LLC Supplier No. A10045
Amyloid Precursor C-Terminal Peptide is a peptide fragment derived from Amyloid Precursor Protein It is designed to model the proteolytic cleavage process of APP which yields -amyloid peptide (A ) associated with amyloid plaques in Alzheimer s disease pathology Amyloid Precursor C-Terminal Peptide influences neuronal functions such as synaptogenesis neural plasticity and iron export regulation Its cleavage product A induces astrocyte proliferation via MAPK and NF- B signaling pathways contributing to neurodegeneration Additionally involvement in inflammation and tissue damage has been reported in animal arthritis models Based on these characteristics Amyloid Precursor C-Terminal Peptide holds research potential in studying A formation neurodegeneration mechanisms and inflammation-associated tissue injury