Learn More
CPC Scientific H-Arg-Gly-Arg-Thr-Ser-Ser-Lys-Glu-Leu-Ala-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific AMYD031A
Encompass
ONE-LETTER SEQUENCE: RGRTSSKELAMOLECULAR FORMULA: C44H81N17O16MOLECULAR WEIGHT:1104.2STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [189125-14-2], RESEARCH AREA: Alzheimer's DiseaseREFERENCES: W.H. van Den Hurk, J. Biol. Chem., 276, 11496 (2001); F.W. van Leeuwen, J. Neural Trans., 57, 137 (1999)
Catalog No. 50-210-8688