Learn More
Diagnostic Biosystems AMYLOID PRECURSOR PROTEIN APP Amyloid Precursor Protein (APP)
Catalog No. 502407519
Encompass_Preferred
This antibody reacts with a 95-100 kDa protein. This antibody recognizes amyloid precursor proteins APP695, APP751 and APP770. Amyloid precursor protein and APP-like proteins are transmembrane glycoproteins with a similar modular domain structure.
Catalog No. 50-240-7519 Supplier Diagnostic Biosystems Supplier No. PDR178
This antibody reacts with a 95-100 kDa protein. This antibody recognizes amyloid precursor proteins APP695, APP751 and APP770. Amyloid precursor protein and APP-like proteins are transmembrane glycoproteins with a similar modular domain structure.