Learn More
Apexbio Technology LLC Amyloid β-Protein (1-15) 183745-81-5 25mg
SDP
Catalog No. 5000092926
Encompass_Preferred
Amyloid -Protein (1-15) (CAS 183745-81-5) is a synthetic peptide fragment corresponding to the amino-terminal residues 1 15 of the amyloid -protein Amyloid -protein (A ) is derived from the amyloid precursor protein (APP) and is a major constituent of amyloid plaques observed in the brains of Alzheimer s disease (AD) patients A peptides aggregate extracellularly disrupting synaptic function and neuronal communication The 1 15 fragment is widely used in studies investigating the molecular mechanisms of A aggregation toxicity antibody binding and its potential role in neurodegenerative pathogenesis This fragment serves as a valuable tool for exploring therapeutic strategies targeting A -driven pathology in AD research
Catalog No. 50-000-92926 Supplier Apexbio Technology LLC Supplier No. A100325
Amyloid -Protein (1-15) (CAS 183745-81-5) is a synthetic peptide fragment corresponding to the amino-terminal residues 1 15 of the amyloid -protein Amyloid -protein (A ) is derived from the amyloid precursor protein (APP) and is a major constituent of amyloid plaques observed in the brains of Alzheimer s disease (AD) patients A peptides aggregate extracellularly disrupting synaptic function and neuronal communication The 1 15 fragment is widely used in studies investigating the molecular mechanisms of A aggregation toxicity antibody binding and its potential role in neurodegenerative pathogenesis This fragment serves as a valuable tool for exploring therapeutic strategies targeting A -driven pathology in AD research