Learn More
Bachem Amyloid Beta Protein (1-40) amide Trifluoroacetate
SDP
Supplier:  Bachem 4095737.0500
Encompass_Preferred
Amyloid Beta Protein (1-40) amide Trifluoroacetate, H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-NH2, 203460-31-5, C194H296N54O57S, Mr 4328.88, 0.5mg. b-Amyloid (1-40), amide.
Catalog No. 50-194-6573