Learn More
Bachem Amyloid Beta Protein (1-42) Trifluoroacetate
SDP
Supplier:  Bachem 4061966.5000
Encompass_Preferred
Amyloid Beta Protein (1-42) Trifluoroacetate, H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH, 107761-42-2, C203H311N55O60S, Mr 4514.10, 5mg.
Catalog No. 50-194-6586