Learn More
Bachem Amyloid Beta Protein (42-1) (HFIP-treated)
SDP
Supplier:  Bachem 4107743.0500
Encompass_Preferred
Amyloid Beta Protein (42-1) (HFIP-treated), H-Ala-Ile-Val-Val-Gly-Gly-Val-Met-Leu-Gly-Ile-Ile-Ala-Gly-Lys-Asn-Ser-Gly-Val-Asp-Glu-Ala-Phe-Phe-Val-Leu-Lys-Gln-His-His-Val-Glu-Tyr-Gly-Ser-Asp-His-Arg-Phe-Glu-Ala-Asp-OH (HFIP-treated), 317366-82-8, C203H311N55O60S, Mr 4514.10, 0.5mg. Reverse sequence of Ab42, inactive control for the Ab. Treating Ab-peptides with 1,1,1,3,3,3-hexafluoroisopropanol (HFIP) is the standard method for disrupting amyloid fibrils and generating the Ab monomers. Working with HFIP-treated Ab-peptides has the advantage of getting more reproducible results whereas untreated Ab-peptides may contain varying amounts and types of oligomeric structures.
Catalog No. NC1561989