Learn More
Amyloid Stain Kit, Congo Red, Each Kit, G-Biosciences

Description
Componets:
- Congo Red Solution: 250 mL
- Hematoxylin Solution: 250 mL
- Bluing Reagent: 250 mL
Specifications
Specifications
| CAS | 573-58-0 |
| Color | Pink to Red |
| Quantity | 3 x 250 mL |
| Molecular Weight (g/mol) | 696.66 |
| Grade | Research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.