Learn More
Scytek Laboratories Amyloid Stain Kit (Congo Red): Large
SDP
Catalog No. NC1969827
Encompass_Preferred
The Amyloid Stain Kit (Congo Red) is intended for use in the histological visualization of amyloid in tissue sections. Examination under a polarizing microscope results in green birefringence of amyloid.Amyloid: Red to PinkErythrocytes: Light OrangeEosinophil Granules: Orange to RedNuclei: BlueKit contents: CRA500, Congo Red Solution, 500 ml.HMM500, Hematoxylin, 500 ml.BRT500, Bluing Reagent, 500 ml.
This item is not returnable. View return policy
Catalog No. NC1969827 Supplier Scytek Laboratories Supplier No. AMY1
The Amyloid Stain Kit (Congo Red) is intended for use in the histological visualization of amyloid in tissue sections. Examination under a polarizing microscope results in green birefringence of amyloid.Amyloid: Red to PinkErythrocytes: Light OrangeEosinophil Granules: Orange to RedNuclei: BlueKit contents: CRA500, Congo Red Solution, 500 ml.HMM500, Hematoxylin, 500 ml.BRT500, Bluing Reagent, 500 ml.