Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amylopectin, MP Biomedicals™

Catalog No. ICN19504883 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19504883 250 g
ICN19504891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19504883 Supplier MP Biomedicals, Inc Supplier No. 0219504883
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9037-22-3
Molecular Formula C30H52O26
Molecular Weight (g/mol) 828.72
MDL Number MFCD00130510
InChI Key WMGFVAGNIYUEEP-WUYNJSITSA-N
Synonym amylopectin, amylopectine, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6s-6-2r,3s,4r,5r,6r-4,5-dihydroxy-3-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6s-6-2r,3s,4r,5r,6r-4,5-dihydroxy-3-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl tetrahydropyran-3-yl oxy-tetrahydropyran-2-yl meth, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-2-2s,3r,4r,5s,6r-3,4-dihydroxy-6-hydroxymethyl-5-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy oxan-2-yl oxy methyl-4,5-dihydroxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 4-o-4-o-alpha-d-glucopyranosyl-6-o-4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439207
ChEBI CHEBI:28057
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6R)-4,5-dihydroxy-3-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-4,5-dihydroxy-2-(hydroxymethy
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OCC3C(C(C(C(O3)OC4C(OC(C(C4O)O)O)CO)O)O)OC5C(C(C(C(O5)CO)O)O)O)CO)O)O)O)O

Specifications

Color White
Physical Form Powder
Quantity 250 g
Chemical Name or Material Amylopectin, from Corn
EINECSNumber 232-911-6
Recommended Storage Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.