Learn More
New England Biolabs, Inc. Amylose Resin High Flow – 100 ml
SDP
Catalog No. 50995070
Encompass
Amylose Resin High Flow is a cross-linked affinity matrix used for the isolation of proteins fused to maltose-binding protein (MBP). This rigid matrix can be used in automated chromatography systems.
Catalog No. 50-995-070 Supplier New England Biolabs, Inc. Supplier No. E8022L
Amylose Resin High Flow is a cross-linked affinity matrix used for the isolation of proteins fused to maltose-binding protein (MBP). This rigid matrix can be used in automated chromatography systems.