Learn More
Medchemexpress LLC β-Amyrin | 559-70-6 | 5 MG
SDP
Catalog No. 5000420437
Encompass_Preferred
β-Amyrin is an orally active natural triterpenoid compound that effectively counteracts amyloid β (Aβ)-induced impairment of long-term potentiation (LTP). It is a promising candidate for Alzheimer's disease (AD) research, exhibiting anti-inflammatory effects, protective activity against pulmonary fibrosis, and notable antibacterial capabilities.
  • Blocks Aβ-induced synaptic dysfunction in hippocampal tissues
  • Shows antioxidant activity
  • Inhibits PGE2, IL-6 secretion, and NF-κB activation
  • Exhibits antibacterial mode of action
  • Ameliorates object recognition memory deficit and neurogenesis impairments
  • Inhibits carrageenan-induced paw edema
  • Protects against pulmonary fibrosis
Catalog No. 50-004-20437 Supplier Medchemexpress LLC Supplier No. HYN29225MG
β-Amyrin is an orally active natural triterpenoid compound that effectively counteracts amyloid β (Aβ)-induced impairment of long-term potentiation (LTP). It is a promising candidate for Alzheimer's disease (AD) research, exhibiting anti-inflammatory effects, protective activity against pulmonary fibrosis, and notable antibacterial capabilities.
  • Blocks Aβ-induced synaptic dysfunction in hippocampal tissues
  • Shows antioxidant activity
  • Inhibits PGE2, IL-6 secretion, and NF-κB activation
  • Exhibits antibacterial mode of action
  • Ameliorates object recognition memory deficit and neurogenesis impairments
  • Inhibits carrageenan-induced paw edema
  • Protects against pulmonary fibrosis