Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-Amyrin | 638-95-9 | 1 MG
SDP

Catalog No. 5000419721
Encompass_Preferred

α-Amyrin is a pentacyclic triterpenoid compound with oral activity. It activates the ERK and GSK-3β signaling pathways, inhibits cancer cell proliferation, and induces apoptosis. It also exhibits anti-bacterial and anti-inflammatory activities and can reduce blood glucose levels. This compound is suitable for research related to cancer, infection, inflammation, metabolic diseases, and neurological diseases, including breast cancer, *Streptococcus oralis* infection, skin inflammation, and diabetes.

  • Purity of 99.99%
  • Molecular weight: 426.72
  • Formula: C30H50O
  • Appearance: Solid, white to off-white
  • Structure classification: Terpenoids, Triterpenes
  • Initial source from plants
  • Store at 4°C, protect from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protect from light
  • Soluble in DMSO (5 mg/mL) and methanol (1 mg/mL)

Catalog No. 50-004-19721 Supplier Medchemexpress LLC Supplier No. HYN84231MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

α-Amyrin is a pentacyclic triterpenoid compound with oral activity. It activates the ERK and GSK-3β signaling pathways, inhibits cancer cell proliferation, and induces apoptosis. It also exhibits anti-bacterial and anti-inflammatory activities and can reduce blood glucose levels. This compound is suitable for research related to cancer, infection, inflammation, metabolic diseases, and neurological diseases, including breast cancer, *Streptococcus oralis* infection, skin inflammation, and diabetes.

  • Purity of 99.99%
  • Molecular weight: 426.72
  • Formula: C30H50O
  • Appearance: Solid, white to off-white
  • Structure classification: Terpenoids, Triterpenes
  • Initial source from plants
  • Store at 4°C, protect from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protect from light
  • Soluble in DMSO (5 mg/mL) and methanol (1 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.